missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)piperidine, 99%, Thermo Scientific Chemicals

Catalog Number 10784094
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5004-07-9
C9H18N2
154.25
MFCD00038011
STWODXDTKGTVCJ-UHFFFAOYSA-N
This item is not returnable. View return policy
5004-07-9
C9H18N2
154.25
MFCD00038011
STWODXDTKGTVCJ-UHFFFAOYSA-N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 52.0°C to 57.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Synonym 4-1-pyrrolidinyl piperidine, 4-pyrrolidin-1-yl piperidine, 4-pyrrolidin-1-yl-piperidine, 4-pyrrolidinopiperidine, 4-1-pyrrolidinyl-piperidine, piperidine, 4-1-pyrrolidinyl, 4-pyrrolidinylpiperidine, 4-pyrrolizinopiperidine, 4-pyrrolidino-piperidine
SMILES C1CCN(C1)C2CCNCC2
IUPAC Name 4-pyrrolidin-1-ylpiperidine
PubChem CID 78703
Formula Weight 154.25
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material 4-(1-Pyrrolidinyl)piperidine
Show More Show Less
compliance-icons
Product Identifier
  • 4-(1-Pyrrolidinyl)piperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'