missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7040207
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
852227-90-8
C16H24BNO2
273.18
MFCD08060504
DWJNNJSONWFVBT-UHFFFAOYSA-N
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid pinacol ester, 4-1-pyrrolidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenylboronic acid pinacol ester, 4-pyrrolidin-1-yl benzeneboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl pyrrolidine
18525715
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCC1
This item is not returnable. View return policy
852227-90-8
C16H24BNO2
273.18
MFCD08060504
DWJNNJSONWFVBT-UHFFFAOYSA-N
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid pinacol ester, 4-1-pyrrolidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenylboronic acid pinacol ester, 4-pyrrolidin-1-yl benzeneboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl pyrrolidine
18525715
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCC1

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 273.18
Percent Purity 97%
Chemical Name or Material 4-(1-Pyrrolidinyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'