missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(1-Piperazinyl)aniline, 97%

Brand:  Alfa Aesar™ H55224.MD

Code : NEW

Additional Details : CAS Number : 67455-41-8 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15451398

  • £15.20 / 250mg


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Description

Description

Applications
4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Reactive with oxidizing agents. Keep at room temperature.
Specifications

Chemical Identifiers

4-(1-Piperazinyl)aniline
177.25
1-(4-Aminophenyl)piperazine
67455-41-8
MFCD01365906

Specifications

4-(1-Piperazinyl)aniline
119° to 123°C
67455-41-8
97%
MFCD01365906
White to purple
250mg
Powder
177.25
1-(4-Aminophenyl)piperazine
Videos
SDS