missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number 11320587
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
916766-81-9
C10H9ClN2O2S
256.704
MFCD09817463
PXFXKTMOZHYJJU-UHFFFAOYSA-N
4-1-methyl-1h-pyrazol-3-yl benzenesulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl-benzenesulfonyl chloride, 4-1-methylpyrazol-3-yl benzenesulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl benzenesulphonyl chloride, chloro 4-1-methylpyrazol-3-yl phenyl sulfone, 3-4-chlorosulphonyl phenyl-1-methyl-1h-pyrazole, 4-1-methyl-1h-pyrazol-3-yl phenylsulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl
24229467
4-(1-methylpyrazol-3-yl)benzenesulfonyl chloride
CN1C=CC(=N1)C2=CC=C(C=C2)S(=O)(=O)Cl
This item is not returnable. View return policy
916766-81-9
C10H9ClN2O2S
256.704
MFCD09817463
PXFXKTMOZHYJJU-UHFFFAOYSA-N
4-1-methyl-1h-pyrazol-3-yl benzenesulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl-benzenesulfonyl chloride, 4-1-methylpyrazol-3-yl benzenesulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl benzenesulphonyl chloride, chloro 4-1-methylpyrazol-3-yl phenyl sulfone, 3-4-chlorosulphonyl phenyl-1-methyl-1h-pyrazole, 4-1-methyl-1h-pyrazol-3-yl phenylsulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl
24229467
4-(1-methylpyrazol-3-yl)benzenesulfonyl chloride
CN1C=CC(=N1)C2=CC=C(C=C2)S(=O)(=O)Cl
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 256.71
Percent Purity 95%
Chemical Name or Material 4-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'