missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(1,2,4-Triazol-1-yl)phenol, 98%

Brand:  Alfa Aesar™ A12109.03

Code : NEW

Additional Details : CAS Number : 68337-15-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11480033

  • £11.30 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

4- (1,2,4-Triazol-1-yl)phenol
C8H7N3O
MFCD00003096
1-(4-Hydroxyphenyl)-1,2,4-triazole
68337-15-5
161.16
608492

Specifications

4- (1,2,4-Triazol-1-yl)phenol
98%
161.16
608492
252° to 257°C
68337-15-5
C8H7N3O
MFCD00003096
1-(4-Hydroxyphenyl)-1,2,4-triazole
1g
Videos
SDS