missing translation for 'onlineSavingsMsg'
Learn More

4-(1,2,3-Thiadiazol-4-yl)benzylamine hydrochloride, ≥95%, Thermo Scientific™

Catalog Number p-2700378
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175205-49-9
C9H10ClN3S
227.71
MFCD02682075
WUUGGOQPXZDQNW-UHFFFAOYSA-N
4-1,2,3-thiadiazol-4-yl benzylamine hydrochloride, 4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 1-4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 4-thiadiazol-4-yl phenyl methanamine hydrochloride, benzenemethanamine,4-1,2,3-thiadiazol-4-yl, 4-1,2,3-thiadiazol-4-yl phenyl methylamine, chloride, 4-4-aminomethyl phenyl-1,2,3-thiadiazole hydrochloride
2737287
[4-(thiadiazol-4-yl)phenyl]methanamine;hydrochloride
C1=CC(=CC=C1CN)C2=CSN=N2.Cl
This item is not returnable. View return policy
175205-49-9
C9H10ClN3S
227.71
MFCD02682075
WUUGGOQPXZDQNW-UHFFFAOYSA-N
4-1,2,3-thiadiazol-4-yl benzylamine hydrochloride, 4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 1-4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 4-thiadiazol-4-yl phenyl methanamine hydrochloride, benzenemethanamine,4-1,2,3-thiadiazol-4-yl, 4-1,2,3-thiadiazol-4-yl phenyl methylamine, chloride, 4-4-aminomethyl phenyl-1,2,3-thiadiazole hydrochloride
2737287
[4-(thiadiazol-4-yl)phenyl]methanamine;hydrochloride
C1=CC(=CC=C1CN)C2=CSN=N2.Cl
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 227.72
Percent Purity 95%
Chemical Name or Material 4-(1, 2, 3-Thiadiazol-4-yl)benzylamine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'