missing translation for 'onlineSavingsMsg'
Learn More

3M™ SecureFit™ Protective Eyewear

Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort

Brand:  3M™ 7100111990

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15282025

  • £4.63 / Each


In Stock 600
Add to basket

Description

Description

  • Need to stock multiple frame sizes at work sites eliminated by self-adjusting feature
  • Secure, snug fit prevents slippage even during motion
  • Anti-fog coating ensures moisture free lenses in hot and humid conditions
  • Anti-scratch coating protects lenses from scratches, nicks and scrapes
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380nm

Specifications

Specifications

Clear
20/Pk.
Polycarbonate
Frameless
Anti-fog
ANSI Z87.1-2010, CSA Z94.3-2007
Proprietary self-adjusting 3M Pressure Diffusion Temple Technology
Videos
Documentation