Learn More
3M™ TR-300 and TR-600 PAPR Battery Packs
Choose from a selection of lithium ion rechargeable batteries for the TR-300 or TR-600 PAPR systems.
Brand: 3M™ 7000144501
239.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.80000kg Transport : UN number : 3480 Chem class : 9
Description
- Rechargeable battery packs
- Lithium Ion
- Long run time for filters and cartridges
Battery packs must be used in accordance with the user instructions, warnings and limitations accompanying each product
Compliance
NIOSH approved.
Specifications
Lithium Ion | |
Battery Pack | |
Versaflo TR-600 PAPR Series | |
<3 hr. | |
0.918 kg |
Blue, Gray | |
Supplying power for PAPR systems | |
TR-630 | |
2 lb. |