missing translation for 'onlineSavingsMsg'
Learn More

3M™ 3M™ Lab Coat

Catalog Number p-6921053
Click to view available options
Size:
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured from SSMMS material. 3M™ Lab Coat has serged seams for durability.

  • Protect against non hazardous dust and liquid splash
  • Knitted cuffs for extra comfort
  • Includes internal pocket on the wearers left hand side
  • Front zip closing
  • Breathable material to help reduce heat build up and promote comfortable wear
  • SSMMS material
Type Lab Coat
Size Large
Gender Unisex
Material SMS
Color Cool White
Closure Type Zipper
Cuff Style Knit
No. of Pockets 1
Collar Type Knit
Seam Type Serged
Certifications/Compliance CE Category 1
For Use With (Application) Protection from non-hazardous liquids and dust
Item Description Lab Coat
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'