Learn More
3M™ Reusable Half Face Mask Respirator, 7500 Series
State-of-the-art features provide unsurpassed comfort. The 3M Reusable Half Face Mask Respirator uses a Bayonet Attachment System for twin lightweight filters to protect against gases, vapors, and particulates.
Brand:  3M™ 7000045950
Code : S8
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10359494
  • £28.10 / Each
In Stock 22
Add to basket
Requires : Compatible cartridges and/or filters. Accommodates 3M 7093 and Series 2000 filters and Series 6000 cartridges.
Description
Description
Durable, reusable and heat-resistant mask is made of advanced silicone material that feels softer on the face, while the unique adjustment design helps reduce tension and pressure points on the face. 
  • Cool Flow™ exhalation valve opens easier, lowering exhalation breathing resistance, reducing heat and moisture buildup in the facepiece
  • Easily adjusted, dual-mode head harness gives user the option of wearing the respirator in either the traditional or drop-down mode
  • Yoke won't get hot like metal yokes, so the 7500 is an excellent choice for welding applications
  • Color-coded sizes for easy identification
 These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Specifications
Specifications
NIOSH 42CFR84
5 x 9.8 x 9.3 in. (12.7 x 24.89 x 23.62cm)
Cartridge 6000 series, filter 2000, 2200, 7000 or 5000 series, or dual airline system
12.7cm
24.89cm
Medium
Bayonet
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
23.62cm
Silicone
Half Mask Respirator
Videos
Documentation