missing translation for 'onlineSavingsMsg'
Learn More

3M™ 4565 Series Disposable Protective Coveralls

Brand:  3M™ 7000034772

Code : Z4

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13467293

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

These key features make this chemical protective coverall a great disposable PPE option. The 3M protective coverall 4565 helps protect against hazardous dusts and light liquid chemical splashes and low pressure sprays. Typical applications may include: Paint Spraying, industrial and pharmaceutical manufacturing, asbestos inspection and handling, and Healthcare. Product recommended for the following industries: Abatement/Remediation, Agriculture, Biological/Biohazard, Cleaning and Maintenance, General Industry/Manufacturing, Painting, Pharmaceutical/Cleanroom.
  • Elastic waist, ankles, and wrists for convenience and freedom of movement.
  • Two-way zipper with storm flap for convenience and to help provide additional protection.
  • Anti-static coating on both sides helps reduce static build-up and prevent the risk of static discharge during use.
  • Does not contain components made from natural rubber latex to help prevent reactions from users with latex sensitivities or restrictions.
  • Does not contain components made from silicone to help prevent reactions from users with silicone sensitivities or restrictions.
Specifications

Specifications

ASTM F1670, EN 14126:2003, EN 1149-1:2006, EN 13034, EN-ISO 13982-30
Polyethylene and Polyester/Polyethylene laminate suit, Metal/Nylon/Polyester Braid zipper, Neoprene Rubber elastic, Polyester/Cotton thread, Polyester cuffs, Polypropylene red tape
91.44 to 99.06cm
Yes
Elastic
White
167 to 176cm
76.2cm
Two-way Zipper with Sealed Storm Flap
Microporous Polyethylene
Elastic
Anti-static
Taped
Medium
Paint spraying, industrial and pharmaceutical manufacturing, asbestos inspection and handling and healthcare
127cm
Videos