missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ White Protective Coverall 4520
95.70 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
95.70 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "22912" to get your promotional price.
95.70 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Designed to offer protection against hazardous dusts and limited liquid splashes and sprays
- Made from lightweight and breathable material for reduced heat stress and premium wearer comfort all day
- Standard collar for use with respiratory equipment or where a hood may not be desired
- Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
Specifications
Size | X-Large |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Cuff Style | Elastic, Knit |
Closure Type | 2-way Zipper with Storm Flap |
Seam Type | Bound |
Coating | Anti-static |
Certifications/Compliance | CE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006 |
Description | White SMMMS Polypropylene suit and green back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'