missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ 4520 Series Disposable Protective Coveralls
Protect against certain light liquid splashes and hazardous dust
Brand: 3M™ 7000034761
Code : 94
Additional Details : Weight : 3.78360kg
Description
- Medium protective overall
- Lightweight
- Breathable giving protection against liquid splash and dust
- Elasticated hood, ankles & waist.
- Two way zip with sealable tab on storm flap
- Three panel hood
- Knitted cuffs
- Low linting
- 3M Catalog# 4520-3XL, 4520-4XL, 4520-L, 4520-M, 4520-XL, 4520-XXL, 4540+3XL
Specifications
4520-M | |
Polypropylene | |
Anti-static | |
Elastic, Knit | |
Yes | |
167 to 176 cm | |
Medium | |
76.2 cm |
2-way Zipper with storm flap | |
CE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006 | |
White | |
White SMMMS Polypropylene suit and green back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread | |
Insulation installation, asbestos and lead abatement, Coal dust in power plants, woodworking, Metal polishing, light-duty building cleaning, general powder handling, general industrial clean-up and maintenance, machine or vehicle maintenance and food processing | |
Bound | |
91.44 to 99.06 cm | |
127 cm |