Learn More
3M™ 4510 Series Disposable Protective Coveralls
The 3M™ Protective Coverall 4510 is disposable safety work wear made of a high quality microporous laminated material. It helps provide basic barrier protection against light liquid splashes and hazardous dusts.
Brand: 3M™ 7000089662
Code : 94
Additional Details : Weight : 4.77000kg
Description
The key features make this protective coverall a great disposable PPE option. The 3M protective coverall 4510 helps protect against light liquid splashes and hazardous dusts. Typical applications for this safety work wear may include: Paint Spraying, Metal Polishing, Machine or Vehicle Maintenance (Oils and Greases), and General Industrial Clean-up and Processing.- Protective coverall helps protect against certain light liquid splashes and hazardous dusts
- Microporous PE laminate offers a barrier against hazardous particles and liquid splashes
- Breathable, SMMMS fabric back panel helps improve air ventilation and helps reduce heat build-up
- Reinforcement gusset between the legs to help provide increased durability during bending and squatting
- Meets government standards for Type 6 Splashes, Type 5 Dust, Anti-Static and Nuclear protection
- Elastic waist and ankles for convenience and freedom of movement.
- Two-way zipper with sealable storm flap for convenience and to help provide additional protection.
- Three panel hood design helps provide a more comfortable fit and a sleek, low-profile style.