missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Hooded Protective Coveralls 4510 Series, Elastic Wrist and Ankles
107.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Type 5/6 Coverall designed to offer cost effective protection against hazardous dusts and limited liquid splashes
- Protection against EN 14126 Biological agents
- Made of a soft lightweight non-woven with microporous laminate for splash protection, strength and breathability
- Certified protection against Type 5/6 hazardous dusts and limited liquid splashes
- 2 panel hood design, 2-way zip with storm flap, elasticated apertures reduce the risk of skin exposure
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
Specifications
Product Type | Cleanroom Coveralls |
Size | 4X-Large |
Material | Microporous Polyethylene Laminate |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Closure Type | 2-way Zipper with Storm Flap |
Seam Type | Bound |
Sterility | Non-sterile |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'