3M™ 4510 Series Disposable Protective Coveralls


Overview
Brand: 3M™

Manufacturer Part Number: 4510WL

UNSPSC: 46181503

Code: 94

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 12962315

Quantity Price
1 £ 106.43 / Pack of 20
Estimated Shipment
12-06-2017
Add to basket


Description and Specification

Specification

Certifications/Compliance EN 13034 Type 6, EN ISO 13982-1 Type 5, EN 1149-1:2006, ASTM D2859-96, CE
Coating Anti-static
Closure Type Two-way Zipper with Storm Flap
Construction White Polypropylene/Polyethylene laminate film suit, Nylon on Polyester Braid zipper, Neoprene Rubber elastic, Polyester thread
Ankle Style Elastic
Cuff Style Elastic
Material Microporous Polyethylene
Seam Type Bound
Color White
Size Large
Disposable Yes
Size (Metric) Chest 99.06 to 109.22cm
Height (Metric) 174 to 181cm
For Use With (Application) Paint spraying, Metal polishing, machine or vehicle maintenance (Oils and greases) and general industrial clean-up and processing, protect against light liquid splashes and hazardous dusts
Latex Free Yes
Size (Metric) Inseam 81.28cm
Size (Metric) Waist 129.54cm