missing translation for 'onlineSavingsMsg'
Learn More

3F8, Tocris Bioscience™

Potent and selective GSK-3β inhibitor

Brand:  Tocris (Bio-Techne) 408310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13464114

  • £165.00 / 10mg


Estimated Shipment
29-10-2018
Add to basket

Description

Description

Potent and selective GSK-3β inhibitor (IC50 values are 34 and 304 nM in the presence of 10 and 100μM ATP respectively).
Specifications

Chemical Identifiers

159109-11-2
286.287
kinome_3660, 5-ethyl-7,8-dimethoxy-1h-pyrrolo 3,4-c isoquinoline-1,3 2h-dione
5-ethyl-7,8-dimethoxypyrrolo[3,4-c]isoquinoline-1,3-dione
C15H14N2O4
ULVWJFBHQIXEPE-UHFFFAOYSA-N
23829003
CCC1=NC2=C(C3=CC(=C(C=C31)OC)OC)C(=O)NC2=O

Specifications

3F8
159109-11-2
kinome_3660, 5-ethyl-7,8-dimethoxy-1h-pyrrolo 3,4-c isoquinoline-1,3 2h-dione
CCC1=NC2=C(C3=CC(=C(C=C31)OC)OC)C(=O)NC2=O
286.287
286.28
10mg
C15H14N2O4
ULVWJFBHQIXEPE-UHFFFAOYSA-N
5-ethyl-7,8-dimethoxypyrrolo[3,4-c]isoquinoline-1,3-dione
23829003
>99%
Videos