missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermo Scientific™ 3D Tank Liner, ASI™ 26 film

Single-use tank liners reduce cleaning steps common to re-usable tanks.

Brand:  Thermo Scientific™ B100373-I

Additional Details : Weight : 7.00000kg

 View more versions of this product

Product Code. 15969786

  • £598.19 / 20 units
Estimated Shipment: 09-03-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.

  • Minimizes bioburden due to construction under sterile cGMP conditions
  • Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications

Specifications

100 L
Individually packed in a header bag
20 per case
Irradiated
Polyethylene
No Tubing
45.7 x 76.2 cm (18 x 30 in.)
HyPerforma™
Room Temperature
ASI™ 26
0 ports
100 L
Videos
SDS
Documents
Certificates
Special Offers

Special Offers