missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3B Scientific™ Life-Size Plastic Skulls
Three-part, life-size replica cast from the skull bones of an adult male.
Brand: 3B Scientific™ A20
Code : 50
Additional Details : Weight : 1.00000kg
Description
- Upper portion of skull has been sectioned horizontally for study of internal structures
- Position and spacing of teeth have been reproduced from a natural denture
- Jaw articulates by means of springs and is detachable
- Skull separates into three parts: calvarium, base of skull and lower jaw
- Dimensions: 7.8 × 5.3 × 6 in. (20 × 13.5 × 15.5cm)
Anatomically Numbered
- Major anatomical details such as bones, fissures and foramina are numbered
- Skull sutures are marked with color for improved clarity
- Includes a key card
With Muscle Attachments
- Muscle attachments (blue) and origins (red) shown in color on the right half
- Skull structures and components on the left half are numbered, totaling more than 140 items
- An excellent demonstration model for teaching purposes
- Includes a key to the number code
Specifications
Anatomical studies of the skull | |
Unbreakable plastic | |
0.6kg |
Detachable skull cap, base of skull and mandible | |
3- part basic anatomical studies of the skull |