missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well, Ultra-Low Binding, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Brand: Corning™ 4588
Additional Details : Weight : 20.60000kg
Description
Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.
Specifications
Standard | |
0.06 cm2 | |
Ultra-Low Attachment | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
20/Pk. | |
11.6 mm |
With Lid | |
16 x 24 | |
Black, Clear Bottom | |
384 | |
Ultra-low Binding | |
100/Cs. | |
127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.) | |
Top/bottom: 3.63/2.82 mm |