384-Well Full Skirted PCR Plates
Reduce well-to-well variability with 384-Well PCR Plate Full Skirted PCR Plates, thin walled for efficient heat transfer and compatible with most major thermal cyclers.
Overview
Brand: Thermo Scientific ABgene
Manufacturer Part Number: TF-0384/W
ABgene 384-well PCR Plate (White) 50 plates
UNSPSC: 12352200
Additional Details:
Additional Details: Weight: 0.99000kg
Product Code. 11579344
Quantity Price
1 £ 183.0 / Pack of 50
Estimated Shipment date
from Supplier 06-03-2017
Add to basket
Description and Specification
Specification
Description ABgene 384-well PCR Plate
Volume (Metric) Working 25µL
Volume (Metric) Well 40µL
Color White
Item Description White; w/Cut-away corner
Certifications/Compliance Certified free of human DNA, RNase, DNase, and endotoxins.
Highlights
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer for equally high performance from every sample
  • SBS footprint
  • 40µL maximum well volume, 25µL working volume
  • Cut Corner: A24