missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 305 dia. x 40mmH Stainless Steel Test Sieve, ASTM Certified, Pore Sizes: Under 10mm

Brand:  RETSCH 60.159.002000

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10064781

  • £175.00 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
40mm
For separation, fractioning, particle size determination
305 mm
2mm
ASTM E 11
Shaker AS 300
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers