missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethoxysilyl)propyl methacrylate, 98%, Thermo Scientific Chemicals

Catalog Number p-3737657
13.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
2530-85-0
C10H20O5Si
248.35
MFCD00008593
XDLMVUHYZWKMMD-UHFFFAOYSA-N
3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
17318
3-trimethoxysilylpropyl 2-methylprop-2-enoate
CC(=C)C(=O)OCCC[Si](OC)(OC)OC
This item is not returnable. View return policy
2530-85-0
C10H20O5Si
248.35
MFCD00008593
XDLMVUHYZWKMMD-UHFFFAOYSA-N
3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
17318
3-trimethoxysilylpropyl 2-methylprop-2-enoate
CC(=C)C(=O)OCCC[Si](OC)(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Density 1.0400g/mL
Boiling Point 190.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2(CH2)3Si(OCH3)3
Refractive Index 1.4300 to 1.4320
Quantity 5 mL
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,ether,methanol and,hydrocarbons
Viscosity 2.5 mm2/s (25°C)
Formula Weight 248.35
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-(Trimethoxysilyl)propyl methacrylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'