missing translation for 'onlineSavingsMsg'
Learn More

3-Thiopheneboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-199840
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6165-69-1
C4H5BO2S
127.95
MFCD00151851
QNMBSXGYAQZCTN-UHFFFAOYSA-N
3-thiopheneboronic acid, thiophene-3-boronic acid, 3-thienylboronic acid, thiophene 3-boronic acid, thiophene-3-boronicacid, thiophen-3-yl-3-boronic acid, 3-thienyl boronic acid, 3-thiophene boronic acid, thiophen-3-yl-boranediol, boronic acid, 3-thienyl
581760
thiophen-3-ylboronic acid
OB(O)C1=CSC=C1
This item is not returnable. View return policy
6165-69-1
C4H5BO2S
127.95
MFCD00151851
QNMBSXGYAQZCTN-UHFFFAOYSA-N
3-thiopheneboronic acid, thiophene-3-boronic acid, 3-thienylboronic acid, thiophene 3-boronic acid, thiophene-3-boronicacid, thiophen-3-yl-3-boronic acid, 3-thienyl boronic acid, 3-thiophene boronic acid, thiophen-3-yl-boranediol, boronic acid, 3-thienyl
581760
thiophen-3-ylboronic acid
OB(O)C1=CSC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 162°C to 167°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 127.96
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Thiopheneboronic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Thiopheneboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'