Learn More
Thermo Scientific Maybridge 3-Thienyl isocyanate, 1g, 95%
Brand: Thermo Scientific Maybridge CC13106DA
3-Thienyl isocyanate, 1g, 95%
Code : 31
Additional Details : CAS Number : 76536-95-3 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC13106DA
3-Thienyl isocyanate, 1g, 95%
Code : 31
Additional Details : CAS Number : 76536-95-3 Weight : 0.05100kg