Learn More
Thermo Scientific Maybridge 3-Thienyl isocyanate, 5g, 95%
Brand: Thermo Scientific Maybridge CC13106DE
3-Thienyl isocyanate, 5g, 95%
Code : 31
Additional Details : CAS Number : 76536-95-3 Weight : 0.05500kg Transport : UN number : 2206 Chem class : 6.1 Pack group : III