missing translation for 'onlineSavingsMsg'
Learn More

(3-Thien-3-ylphenyl)methanol, ≥97%, Thermo Scientific™

Catalog Number p-3176117
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
89929-82-8
C11H10OS
190.26
MFCD06203083
VSWBZMMHNMDWJL-UHFFFAOYSA-N
3-thien-3-ylphenyl methanol, 3-thiophen-3-yl phenyl methanol, 3-3-thienyl phenyl methanol, 3-thiophen-3-ylphenyl methanol, acmc-20ls08, 3-3-thienyl phenylmethanol, 3-3-thiophenyl phenyl methanol, 3-3-thienyl phenyl methan-1-ol
7164662
(3-thiophen-3-ylphenyl)methanol
C1=CC(=CC(=C1)CO)C2=CSC=C2
This item is not returnable. View return policy
89929-82-8
C11H10OS
190.26
MFCD06203083
VSWBZMMHNMDWJL-UHFFFAOYSA-N
3-thien-3-ylphenyl methanol, 3-thiophen-3-yl phenyl methanol, 3-3-thienyl phenyl methanol, 3-thiophen-3-ylphenyl methanol, acmc-20ls08, 3-3-thienyl phenylmethanol, 3-3-thiophenyl phenyl methanol, 3-3-thienyl phenyl methan-1-ol
7164662
(3-thiophen-3-ylphenyl)methanol
C1=CC(=CC(=C1)CO)C2=CSC=C2
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 190.27
Percent Purity 95%
Chemical Name or Material (3-Thien-3-ylphenyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'