missing translation for 'onlineSavingsMsg'
Learn More

3-[(Tetrahydrofuran-2-ylmethyl)amino]propanenitrile, 97%, Thermo Scientific™

Catalog Number p-3830083
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
90322-18-2
C8H14N2O
154.21
MFCD00121811
ZSKZHSOMHSHFDF-UHFFFAOYNA-N
3-tetrahydrofuran-2-ylmethyl amino propanenitrile, 3-oxolan-2-ylmethyl amino propanenitrile, 3-tetrahydro-2-furanylmethyl amino-propanenitrile, 3-tetrahydrofuran-2-yl methyl amino propanenitrile, 3-oxolan-2-ylmethylamino propanenitrile, 3-oxolan-2-yl methyl amino propanenitrile, 3-tetrahydro-furan-2-ylmethyl-amino-propionitrile, 3-tetrahydrofuran-2-yl ;methyl ;amino ;propanenitrile, propanenitrile,3-tetrahydro-2-furanyl methyl amino
2779328
3-(oxolan-2-ylmethylamino)propanenitrile
N#CCCNCC1CCCO1
This item is not returnable. View return policy
90322-18-2
C8H14N2O
154.21
MFCD00121811
ZSKZHSOMHSHFDF-UHFFFAOYNA-N
3-tetrahydrofuran-2-ylmethyl amino propanenitrile, 3-oxolan-2-ylmethyl amino propanenitrile, 3-tetrahydro-2-furanylmethyl amino-propanenitrile, 3-tetrahydrofuran-2-yl methyl amino propanenitrile, 3-oxolan-2-ylmethylamino propanenitrile, 3-oxolan-2-yl methyl amino propanenitrile, 3-tetrahydro-furan-2-ylmethyl-amino-propionitrile, 3-tetrahydrofuran-2-yl ;methyl ;amino ;propanenitrile, propanenitrile,3-tetrahydro-2-furanyl methyl amino
2779328
3-(oxolan-2-ylmethylamino)propanenitrile
N#CCCNCC1CCCO1
CAS Min % 97.0
Assay Percent Range 97%
Formula Weight 154.21
Percent Purity 97%
Quantity 1 g
Packaging Amber Glass Bottle
Chemical Name or Material 3-[(Tetrahydrofuran-2-ylmethyl)amino]propanenitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'