missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butylphenol, 99+%, Thermo Scientific Chemicals

Catalog Number p-80835
Click to view available options
Quantity:
1 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
585-34-2
C10H14O
150.22
MFCD00002300
CYEKUDPFXBLGHH-UHFFFAOYSA-N
m-tert-butylphenol, phenol, 3-1,1-dimethylethyl, 3-t-butylphenol, 3-tert-butyl phenol, phenol, m-tert-butyl, 3-tert-butyl-phenol, 3-tertbutylphenol, 3-tert.butylphenol, m-tert.-butylphenol
11450
CHEBI:34348
3-tert-butylphenol
CC(C)(C)C1=CC=CC(O)=C1
This item is not returnable. View return policy
585-34-2
C10H14O
150.22
MFCD00002300
CYEKUDPFXBLGHH-UHFFFAOYSA-N
m-tert-butylphenol, phenol, 3-1,1-dimethylethyl, 3-t-butylphenol, 3-tert-butyl phenol, phenol, m-tert-butyl, 3-tert-butyl-phenol, 3-tertbutylphenol, 3-tert.butylphenol, m-tert.-butylphenol
11450
CHEBI:34348
3-tert-butylphenol
CC(C)(C)C1=CC=CC(O)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 40°C to 43°C
Color White
Boiling Point 125°C to 130°C (20.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4OH
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 150.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3-tert-Butylphenol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-tert-Butylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'