missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-tert-Butyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63410.03

Code : NEW

Additional Details : CAS Number : 934168-94-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15440089

  • £119.78 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

3-tert-Butyl-2-thiohydantoin
172.25
15479284
934168-94-2
MFCD14061126
3-tert-Butyl-2-thioxo-4-imidazolidinone

Specifications

3-tert-Butyl-2-thiohydantoin
Solid
172.25
15479284
1g
934168-94-2
95%
MFCD14061126
3-tert-Butyl-2-thioxo-4-imidazolidinone
Videos
SDS