missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyl)-1H-pyrazol-5-amine, 97%, Maybridge

Brand:  Maybridge KM00316EB

Code : 31

Additional Details : CAS Number : 82560-12-1 Weight : 0.07500kg

Quantity 25g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10182754

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3-(tert-Butyl)-1H-pyrazol-5-amine
97.0
97%
139.2
82560-12-1
97%
C7H13N3
MFCD00067987

Specifications

3-(tert-Butyl)-1H-pyrazol-5-amine
97.0
97%
Amber Glass Bottle
MFCD00067987
82560-12-1
97%
C7H13N3
139.2
25g
Videos
SDS