Learn More
Thermo Scientific Maybridge 3-(tert-Butyl)-1H-pyrazol-5-amine, 1g, 97%
Brand: Thermo Scientific Maybridge KM00316DA
3-(tert-Butyl)-1H-pyrazol-5-amine, 1g, 97%
Code : 31
Additional Details : CAS Number : 82560-12-1 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge KM00316DA
3-(tert-Butyl)-1H-pyrazol-5-amine, 1g, 97%
Code : 31
Additional Details : CAS Number : 82560-12-1 Weight : 0.05100kg