missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-tert-Butyl-1-methyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H64000.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15451009

  • £91.90 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

3-tert-Butyl-1-methyl-2-thiohydantoin
186.27
MFCD27976893
3-tert-Butyl-1-methyl-2-thioxo-4-imidazolidinone

Specifications

3-tert-Butyl-1-methyl-2-thiohydantoin
95%
MFCD27976893
1g
Solid
186.27
3-tert-Butyl-1-methyl-2-thioxo-4-imidazolidinone
Videos
SDS