missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Pyrrolidin-1-ylphenyl)methylamine, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 176.26 |
Percent Purity | 90% |
Chemical Name or Material | (3-Pyrrolidin-1-ylphenyl)methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'