missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-5-ylbenzoic acid, ≥97%, Thermo Scientific™

Catalog Number p-3110490
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
852180-74-6
C11H8N2O2
200.197
MFCD05864810
URQRFVRGUHUTMI-UHFFFAOYSA-N
3-pyrimidin-5-yl-benzoic acid, 3-pyrimidin-5-yl benzoic acid, 3-5-pyrimidinyl benzoic acid, benzoic acid,3-5-pyrimidinyl, benzoic acid, 3-5-pyrimidinyl, 3-pyrimidin-5-yl-benzoicacid, benzoic acid, 3-5-pyrimidinyl-9ci
4187250
3-pyrimidin-5-ylbenzoic acid
C1=CC(=CC(=C1)C(=O)O)C2=CN=CN=C2
This item is not returnable. View return policy
852180-74-6
C11H8N2O2
200.197
MFCD05864810
URQRFVRGUHUTMI-UHFFFAOYSA-N
3-pyrimidin-5-yl-benzoic acid, 3-pyrimidin-5-yl benzoic acid, 3-5-pyrimidinyl benzoic acid, benzoic acid,3-5-pyrimidinyl, benzoic acid, 3-5-pyrimidinyl, 3-pyrimidin-5-yl-benzoicacid, benzoic acid, 3-5-pyrimidinyl-9ci
4187250
3-pyrimidin-5-ylbenzoic acid
C1=CC(=CC(=C1)C(=O)O)C2=CN=CN=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 200.2
Percent Purity 97%
Chemical Name or Material 3-Pyrimidin-5-ylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'