missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-5-ylbenzaldehyde, 97%, Thermo Scientific™

Catalog Number 10525953
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
640769-70-6
C11H8N2O
184.198
MFCD05864833
RAHJVJBPRWJPHE-UHFFFAOYSA-N
3-pyrimidin-5-yl benzaldehyde, 3-pyrimidin-5-yl-benzaldehyde, 3-5-pyrimidinyl benzaldehyde, benzaldehyde, 3-5-pyrimidinyl, acmc-1b2vh, benzaldehyde,3-5-pyrimidinyl
4187249
3-pyrimidin-5-ylbenzaldehyde
C1=CC(=CC(=C1)C=O)C2=CN=CN=C2
This item is not returnable. View return policy
640769-70-6
C11H8N2O
184.198
MFCD05864833
RAHJVJBPRWJPHE-UHFFFAOYSA-N
3-pyrimidin-5-yl benzaldehyde, 3-pyrimidin-5-yl-benzaldehyde, 3-5-pyrimidinyl benzaldehyde, benzaldehyde, 3-5-pyrimidinyl, acmc-1b2vh, benzaldehyde,3-5-pyrimidinyl
4187249
3-pyrimidin-5-ylbenzaldehyde
C1=CC(=CC(=C1)C=O)C2=CN=CN=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 184.2
Percent Purity 97%
Chemical Name or Material 3-Pyrimidin-5-ylbenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'