missing translation for 'onlineSavingsMsg'
Learn More

(3-Pyrimidin-2-ylphenyl)methanol, 97%, Thermo Scientific™

Catalog Number p-3527228
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
892502-12-4
C11H10N2O
186.21
MFCD09064977
WMNXGJMCXOLBBV-UHFFFAOYSA-N
3-pyrimidin-2-ylphenyl methanol, 3-pyrimidin-2-yl phenyl methanol, benzenemethanol,3-2-pyrimidinyl, 3-pyrimidin-2-yl-benzyl alcohol, benzenemethanol, 3-2-pyrimidinyl, 3-pyrimidin-2-ylphenyl methan-1-ol
24229565
OCC1=CC=CC(=C1)C1=NC=CC=N1
This item is not returnable. View return policy
892502-12-4
C11H10N2O
186.21
MFCD09064977
WMNXGJMCXOLBBV-UHFFFAOYSA-N
3-pyrimidin-2-ylphenyl methanol, 3-pyrimidin-2-yl phenyl methanol, benzenemethanol,3-2-pyrimidinyl, 3-pyrimidin-2-yl-benzyl alcohol, benzenemethanol, 3-2-pyrimidinyl, 3-pyrimidin-2-ylphenyl methan-1-ol
24229565
OCC1=CC=CC(=C1)C1=NC=CC=N1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name (3-pyrimidin-2-ylphenyl)methanol
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material (3-Pyrimidin-2-ylphenyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'