missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pyrimidin-2-ylbenzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 3-pyrimidin-2-yl-benzoic acid, 3-pyrimidin-2-yl benzoic acid, 2-3-carboxyphenyl pyrimidine, 3-2-pyrimidinyl benzoic acid, benzoic acid,3-2-pyrimidinyl, benzoic acid, 3-2-pyrimidinyl, 3-pyrimidin-2-yl benzoicacid |
Formula Weight | 200.2 |
Percent Purity | 97% |
Chemical Name or Material | 3-Pyrimidin-2-ylbenzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'