missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridyl isothiocyanate, ≥95%, Thermo Scientific™

Catalog Number p-2700310
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
17452-27-6
C6H4N2S
136.172
MFCD00052351
VMSZFBSYWXMXRF-UHFFFAOYSA-N
3-pyridyl isothiocyanate, pyridine, 3-isothiocyanato, 3-isothiocyanato-pyridine, pyridine-3-isothiocyanate, pyridine,3-isothiocyanato, 3-pyridylisothiocyanate, 3-pyridyl isotiocyanate, acmc-20an1z, 3-pyridyl-isothiocyanate, pyridin-3-isothiocyanate
2737242
3-isothiocyanatopyridine
C1=CC(=CN=C1)N=C=S
This item is not returnable. View return policy
17452-27-6
C6H4N2S
136.172
MFCD00052351
VMSZFBSYWXMXRF-UHFFFAOYSA-N
3-pyridyl isothiocyanate, pyridine, 3-isothiocyanato, 3-isothiocyanato-pyridine, pyridine-3-isothiocyanate, pyridine,3-isothiocyanato, 3-pyridylisothiocyanate, 3-pyridyl isotiocyanate, acmc-20an1z, 3-pyridyl-isothiocyanate, pyridin-3-isothiocyanate
2737242
3-isothiocyanatopyridine
C1=CC(=CN=C1)N=C=S
CAS Min % 94.5
CAS Max % 100.0
Density 1.22g/mL
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 136.18
Percent Purity 95%
Chemical Name or Material 3-Pyridyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'