missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridinealdoxime, 98%, ACROS Organics™

Catalog Number p-189448
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1193-92-6
C6H6N2O
122.13
MFCD00006409,MFCD00006409
YBKOPFQCLSPTPV-VMPITWQZSA-N
nicotinaldoxime, 3-pyridinealdoxime, nicotinaldehyde oxime, pyridine-3-aldoxime, 3-pyridinecarboxaldehyde, oxime, nicotinaldehyde, oxime, 3-pyridine-aldoxime, pyridine, 3-formyl-, oxime, hydroxyimino-3-pyridylmethane, pyridinaldoxim
5371219
(E)-N-[(pyridin-3-yl)methylidene]hydroxylamine
O\N=C\C1=CC=CN=C1
This item is not returnable. View return policy
1193-92-6
C6H6N2O
122.13
MFCD00006409,MFCD00006409
YBKOPFQCLSPTPV-VMPITWQZSA-N
nicotinaldoxime, 3-pyridinealdoxime, nicotinaldehyde oxime, pyridine-3-aldoxime, 3-pyridinecarboxaldehyde, oxime, nicotinaldehyde, oxime, 3-pyridine-aldoxime, pyridine, 3-formyl-, oxime, hydroxyimino-3-pyridylmethane, pyridinaldoxim
5371219
(E)-N-[(pyridin-3-yl)methylidene]hydroxylamine
O\N=C\C1=CC=CN=C1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 149°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 122.13
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Pyridinealdoxime, 98%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'