missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Maybridge 3-Pyridin-3-ylaniline, 5g, 97%

Brand:  Thermo Scientific Maybridge CC39814DE

3-Pyridin-3-ylaniline, 5g, 97%

Code : 31

Additional Details : CAS Number : 57976-57-5 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Product Code. 10235484

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.