missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pyridin-3-ylaniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 170.21 |
Percent Purity | 97% |
Chemical Name or Material | 3-Pyridin-3-ylaniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'