missing translation for 'onlineSavingsMsg'
Learn More

(3-Pyrid-4-ylphenyl)methanol, 97%, Thermo Scientific™

Catalog Number p-3109891
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
85553-55-5
C12H11NO
185.23
MFCD06659083
ZORHPGKUTIFODF-UHFFFAOYSA-N
2795564
(3-pyridin-4-ylphenyl)methanol
OCC1=CC=CC(=C1)C1=CC=NC=C1
This item is not returnable. View return policy
85553-55-5
C12H11NO
185.23
MFCD06659083
ZORHPGKUTIFODF-UHFFFAOYSA-N
2795564
(3-pyridin-4-ylphenyl)methanol
OCC1=CC=CC(=C1)C1=CC=NC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-pyrid-4-ylphenyl methanol, 3-pyridin-4-yl phenyl methanol, 3-pyridin-4-yl benzyl alcohol, benzenemethanol,3-4-pyridinyl, 3-pyridin-4-ylphenyl methanol, 3-4-pyridyl phenyl methan-1-ol, benzenemethanol, 3-4-pyridinyl, 4-3-hydroxymethyl phenyl pyridine
Formula Weight 185.22
Percent Purity 97%
Chemical Name or Material (3-Pyrid-4-ylphenyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'