missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyrid-2-yloxy)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-3485356
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
51362-30-2
C12H9NO3
215.208
MFCD09025853
LYSIEAIIZBZRCE-UHFFFAOYSA-N
3-pyridin-2-yloxy benzoic acid, 3-pyrid-2-yloxy benzoic acid, 3-2-pyridyloxy benzoic acid, 3-2 pyridyloxy-benzoic acid, 3-pyridin-2-yl oxy benzoic acid, benzoic acid, 3-2-pyridinyloxy
21901423
3-pyridin-2-yloxybenzoic acid
C1=CC=NC(=C1)OC2=CC=CC(=C2)C(=O)O
This item is not returnable. View return policy
51362-30-2
C12H9NO3
215.208
MFCD09025853
LYSIEAIIZBZRCE-UHFFFAOYSA-N
3-pyridin-2-yloxy benzoic acid, 3-pyrid-2-yloxy benzoic acid, 3-2-pyridyloxy benzoic acid, 3-2 pyridyloxy-benzoic acid, 3-pyridin-2-yl oxy benzoic acid, benzoic acid, 3-2-pyridinyloxy
21901423
3-pyridin-2-yloxybenzoic acid
C1=CC=NC(=C1)OC2=CC=CC(=C2)C(=O)O
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 215.21
Percent Purity 95%
Chemical Name or Material 3-(Pyrid-2-yloxy)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'