missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinyl(1-pyrrolidinyl)methanone, 97%, Thermo Scientific™

Catalog Number p-3075514
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
35090-94-9
C10H18N2O
182.267
MFCD05865122
AMOUVOLDCHVMBJ-UHFFFAOYSA-N
piperidin-3-yl pyrrolidin-1-yl methanone, 3-piperidinyl 1-pyrrolidinyl methanone, 3-pyrrolidin-1-ylcarbonyl piperidine, 3-pyrrolidine-1-carbonyl piperidine, 3-piperidyl 1-pyrrolidyl methanone, piperidin-3-yl-pyrrolidin-1-yl-methanone, pyrrolidine, 1-3-piperidinylcarbonyl, r-piperidin-3-yl pyrrolidin-1-yl methanone, 3-piperidyl pyrrolidinyl ketone, 3r-3-pyrrolidin-1-ylcarbonyl piperidine
2794682
piperidin-3-yl(pyrrolidin-1-yl)methanone
C1CCN(C1)C(=O)C2CCCNC2
This item is not returnable. View return policy
35090-94-9
C10H18N2O
182.267
MFCD05865122
AMOUVOLDCHVMBJ-UHFFFAOYSA-N
piperidin-3-yl pyrrolidin-1-yl methanone, 3-piperidinyl 1-pyrrolidinyl methanone, 3-pyrrolidin-1-ylcarbonyl piperidine, 3-pyrrolidine-1-carbonyl piperidine, 3-piperidyl 1-pyrrolidyl methanone, piperidin-3-yl-pyrrolidin-1-yl-methanone, pyrrolidine, 1-3-piperidinylcarbonyl, r-piperidin-3-yl pyrrolidin-1-yl methanone, 3-piperidyl pyrrolidinyl ketone, 3r-3-pyrrolidin-1-ylcarbonyl piperidine
2794682
piperidin-3-yl(pyrrolidin-1-yl)methanone
C1CCN(C1)C(=O)C2CCCNC2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Formula Weight 182.27
Percent Purity 97%
Quantity 250 mg
Chemical Name or Material 3-Piperidinyl(1-pyrrolidinyl)methanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'