missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Piperidinyl(1-pyrrolidinyl)methanone, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Formula Weight | 182.27 |
Percent Purity | 97% |
Quantity | 5 g |
Chemical Name or Material | 3-Piperidinyl(1-pyrrolidinyl)methanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'