Learn More
Thermo Scientific Maybridge (3-Piperidinophenyl)methylamine, 1g, 95%
Brand: Thermo Scientific Maybridge CC34013DA
(3-Piperidinophenyl)methylamine, 1g, 95%
Code : 31
Additional Details : Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III