Learn More
Thermo Scientific Maybridge (3-Piperidinophenyl)methylamine, 5g, 95%
Brand: Thermo Scientific Maybridge CC34013DE
(3-Piperidinophenyl)methylamine, 5g, 95%
Code : 31
Additional Details : CAS Number : 175696-71-6 Weight : 0.05500kg Transport : UN number : 2735 Chem class : 8 Pack group : III