missing translation for 'onlineSavingsMsg'
Learn More

3-(Piperidinomethyl)benzaldehyde, ≥97%, Thermo Scientific™

Catalog Number p-3175967
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
471930-01-5
C13H17NO
203.285
MFCD07782418
UJDTVJKIOWAUNQ-UHFFFAOYSA-N
3-piperidinomethyl benzaldehyde, 3-piperidin-1-ylmethyl benzaldehyde, 3-piperidylmethyl benzaldehyde, 3-1-piperidinylmethyl benzaldehyde, 3-1-piperidinyl methylbenzaldehyde, 1-3-carboxaldehydebenzyl piperidine
7164643
3-(piperidin-1-ylmethyl)benzaldehyde
C1CCN(CC1)CC2=CC=CC(=C2)C=O
This item is not returnable. View return policy
471930-01-5
C13H17NO
203.285
MFCD07782418
UJDTVJKIOWAUNQ-UHFFFAOYSA-N
3-piperidinomethyl benzaldehyde, 3-piperidin-1-ylmethyl benzaldehyde, 3-piperidylmethyl benzaldehyde, 3-1-piperidinylmethyl benzaldehyde, 3-1-piperidinyl methylbenzaldehyde, 1-3-carboxaldehydebenzyl piperidine
7164643
3-(piperidin-1-ylmethyl)benzaldehyde
C1CCN(CC1)CC2=CC=CC(=C2)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 203.28
Percent Purity 97%
Chemical Name or Material 3-(Piperidinomethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'