missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinobenzoic acid, 97%, Thermo Scientific™

Catalog Number p-3110469
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
77940-94-4
C12H15NO2
205.257
MFCD06659081
FLQRORYAJSTYLT-UHFFFAOYSA-N
3-piperidinobenzoic acid, 3-piperidin-1-yl benzoic acid, 3-piperidin-1-yl-benzoic acid, 1-3-carboxyphenyl piperidine, 3-1-piperidinyl benzoic acid, 3-piperidylbenzoic acid, benzoic acid,3-1-piperidinyl, benzoic acid, 3-1-piperidinyl
2795552
3-piperidin-1-ylbenzoic acid
C1CCN(CC1)C2=CC=CC(=C2)C(=O)O
This item is not returnable. View return policy
77940-94-4
C12H15NO2
205.257
MFCD06659081
FLQRORYAJSTYLT-UHFFFAOYSA-N
3-piperidinobenzoic acid, 3-piperidin-1-yl benzoic acid, 3-piperidin-1-yl-benzoic acid, 1-3-carboxyphenyl piperidine, 3-1-piperidinyl benzoic acid, 3-piperidylbenzoic acid, benzoic acid,3-1-piperidinyl, benzoic acid, 3-1-piperidinyl
2795552
3-piperidin-1-ylbenzoic acid
C1CCN(CC1)C2=CC=CC(=C2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 205.26
Percent Purity 97%
Chemical Name or Material 3-Piperidinobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'